EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
GPR109a agonists. Part 1: 5-Alkyl and 5-aryl-pyrazole-tetrazoles as agonists of the human orphan G-protein coupled receptor GPR109a.
PMID 19307116
Abstract
5-Alkyl and aryl-pyrazole-tetrazoles have been identified as a new class of selective, small-molecule, agonists of the human G-protein-coupled receptor GPR109a, a high affinity receptor for the HDL-raising drug nicotinic acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
524697
2,3,5-Trifluorophenylboronic acid, ≥95%
C6H4BF3O2