EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Interactive effects of sulfur and nitrogen supply on the concentration of sinigrin and allyl isothiocyanate in Indian mustard (Brassica juncea L.).
PMID 19309148
Abstract
Food derived from Brassica species is rich in glucosinolates. Hydrolysis of these compounds by myrosinase yields isothiocyanates and other breakdown products, which due to their pungency represent the primary purpose of Indian mustard cultivation. Strong interactive effects of S (0.0, 0.2, and 0.6 g pot(-1)) and N (1, 2, and 4 g pot(-1)) supply on growth, seed yield, and the concentrations of glucosinolates and isothiocyanates in seeds were observed in growth experiments, reflecting the involvement of S-containing amino acids in both protein and glucosinolate synthesis. At intermediate S supply, a strong N-induced S limitation was apparent, resulting in high concentrations of sinigrin (12 micromol g(-1) of DM) and allyl isothiocyanate (213 micromol kg(-1) of DM) at low N supply only. Myrosinase activity in seeds increased under low N and low S supply, but the results do not suggest that sinigrin functions as a transient reservoir for S.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1647
(−)-Sinigrin hydrate, from horseradish
C10H16KNO9S2 · xH2O
00290
(−)-Sinigrin hydrate, analytical standard
C10H16KNO9S2 · xH2O
85440
(−)-Sinigrin hydrate, ≥99.0% (TLC)
C10H16KNO9S2 · xH2O