EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Palladium-catalyzed intramolecular direct arylation of benzoic acids by tandem decarboxylation/C-H activation.
PMID 19317495
Abstract
A novel protocol for the palladium-catalyzed intramolecular direct arylation of benzoic acids is reported, which combines decarboxylation and C-H activation into one efficient process. With the optimized catalyst system of palladium trifluoroacetate, silver carbonate, and the proper solvent mixture (5% DMSO/dioxane), dibenzofuran derivatives are formed in excellent yield and selectivity.