EMAIL THIS PAGE TO A FRIEND
Neuron
Selective gating of glutamatergic inputs to excitatory neurons of amygdala by presynaptic GABAb receptor.
PMID 19324000
Abstract
GABAb receptor (GABAbR)-mediated suppression of glutamate release is critical for limiting glutamatergic transmission across the central nervous system (CNS). Here we show that, upon tetanic stimulation of afferents to lateral amygdala, presynaptic GABAbR-mediated inhibition only occurs in glutamatergic inputs to principle neurons (PNs), not to interneurons (INs), despite the presence of GABAbR in terminals to both types of neurons. The selectivity is caused by differential local GABA accumulation; it requires GABA reuptake and parallels distinct spatial distributions of presynaptic GABAbR in terminals to PNs and INs. Moreover, GABAbR-mediated suppression of theta-burst-induced long-term potentiation (LTP) occurs only in the inputs to PNs, not to INs. Thus, target-cell-specific control of glutamate release by presynaptic GABAbR orchestrates the inhibitory dominance inside amygdala and might contribute to prevention of nonadaptive defensive behaviors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0593 CGP52432, ≥98% (HPLC)
C15H24Cl2NO4P
S9066
SKF-89976A, >98% (HPLC), solid
C22H25NO2 · HCl