EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Grandine A, a new proaporphine alkaloid from the bark of Phoebe grandis.
PMID 19325519
Abstract
The stem bark of Phoebe grandis afforded one new oxoproaporphine; (-)-grandine A (1), along with six known isoquinoline alkaloids: (-)-8,9-dihydrolinearisine (2), boldine, norboldine, lauformine, scortechiniine A and scortechiniine B. In addition to that of the new compound, complete 1H- and 13C-NMR data of the tetrahydroproaporphine (-)-8,9-dihydrolinearisine (2) is also reported. The alkaloids' structures were elucidated primarily by means of high field 1D- and 2D-NMR and HRMS spectral data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3916
Boldine, analytical standard
C19H21NO4