EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Phosphorus trichloride-mediated and microwave-assisted synthesis of a small collection of amides bearing strong electron-withdrawing group substituted anilines.
PMID 19326911