EMAIL THIS PAGE TO A FRIEND
Skin therapy letter
Rosacea and its topical management.
PMID 19330270
Abstract
Many options exist for the treatment of rosacea, including topical and systemic therapies, laser and light-based therapies, and surgical procedures. A classification system for rosacea identifies 4 subtypes (i.e., erythematotelangiectatic, papulopustular, phymatous, and ocular), which may help guide therapeutic decision-making. The goals of therapy include reduction of papules, pustules, erythema, physical discomfort, and an improvement in quality of life. Standard topical treatment agents include metronidazole, azelaic acid, and sodium sulfacetamide-sulfur. Second line therapies include benzoyl peroxide, clindamycin, calcineurin inhibitors, and permethrin.
Related Materials