EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Fluorescent chemosensor for reactive organohalides in micellar solution with an example of autocatalysis.
PMID 19333457
Abstract
N-Alkylation of a fluorescent macrocyclic amine in aqueous micellar solution produces enhanced emission; the reaction with chloromethyl methyl ether exhibits autocatalysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100331
Chloromethyl methyl ether, technical grade
C2H5ClO