EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Fate of benfuracarb insecticide in mollisols and brinjal crop.
PMID 19333533
Abstract
The fate of benfuracarb was studied under field conditions in brinjal fruits and soil following foliar spray application at 0.25 and 0.50 microg g(-1) by HPLC. At 0.25 microg g(-1), benfuracarb persisted up to 7 days both in soil and brinjal but at 0.50 microg g(-1), benfuracarb residues persisted up to 10 and 12 days in soil and brinjal fruits, respectively. The persistence of benfuracarb residues, both in soil and brinjal, followed first-order kinetics. The half-life values of benfuracarb in soil and brinjal fruit were found to be 3.54 and 3.90 days at 0.25 microg g(-1) and 3.75 and 4.73 days at 0.50 microg g(-1), respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31544
Benfuracarb, PESTANAL®, analytical standard
C20H30N2O5S