EMAIL THIS PAGE TO A FRIEND
Cancer prevention research (Philadelphia, Pa.)
Liposomal encapsulation of deguelin: evidence for enhanced antitumor activity in tobacco carcinogen-induced and oncogenic K-ras-induced lung tumorigenesis.
PMID 19336726
Abstract
Deguelin has shown promising chemopreventive and therapeutic activities in diverse types of cancers. However, the potential side effect of deguelin over a certain dose could be the substantial hurdle in the practical application of the drug. One of the successful strategies for the use of deguelin in clinical trials could be lung-specific delivery of the drug. The present study evaluates the efficacy of liposome-encapsulated deguelin with a dose of 0.4 mg/kg, which is 10 times less than the dose (4 mg/kg) for preventive and therapeutic activities validated in previous in vivo studies. Liposomal deguelin revealed cytotoxic activity in vitro in premalignant and malignant human bronchial epithelial cells and non-small cell lung cancer cells through the same mechanistic pathway previously reported for deguelin (i.e., suppression of the heat shock protein 90 chaperone function and induction of apoptosis). Delivery of liposomal deguelin at a dose of 0.4 mg/kg by intranasal instillation resulted in markedly increased drug partitioning to the lungs compared with that of 4 mg/kg deguelin or 0.4 mg/kg liposomal deguelin administered by oral gavage. Lung-specific delivery of deguelin (0.4 mg/kg) via nasal or intratracheal instillation in a liposomal formulation also showed significant chemopreventive and therapeutic activities in 4-(methylnitrosoamino)-1-(3-pyridyl)-1-butanone/benzo(a)pyrene-treated A/J mice and K-rasLAC57Bl6/129/sv F1 mice with no detectable toxicity. Our findings support the potential use of deguelin in a liposomal formulation via lung-specific delivery to improve efficacy and to reduce the potential side effects of the agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0817
(−)-Deguelin, >98% (HPLC), powder
C23H22O6