EMAIL THIS PAGE TO A FRIEND
The Canadian veterinary journal = La revue veterinaire canadienne
Chronic intermittent diarrhea in a 14-month-old Abyssinian cat.
PMID 19337620
Abstract
A 14-month-old intact, female Abyssinian cat was presented for chronic intermittent diarrhea and bilateral enlargement of the mammary glands. Gastrointestinal coccidiosis was diagnosed; therapy with sulfadi-methoxine was unsuccessful in the elimination of Isospora felis and clinical signs. Infection with Tritrichomonas foetus was diagnosed by fecal polymerase chain reaction (PCR) and successfully treated with ronidazole and dietary modification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7635
Ronidazole, analytical standard
C6H8N4O4