EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
The aqueous photosensitized degradation of butylparaben.
PMID 19337670
Abstract
The photosensitized oxidation of butyl p-hydroxybenzoate (butylparaben, BP), the endocrine disrupting compound, in aqueous solution using rose bengal (RB) and aluminium phthalocyanine chloride tetrasulfonic acid (PC) as sensitizers was examined. A xenon lamp simulating solar radiation was used as a light source. The influence of pH of the reaction mixture, oxygen content, irradiation intensity and initial sensitizers and BP concentration on the reaction rate was studied. Based on the kinetic model, the rate constants of singlet oxygen quenching and reaction with BP as well as the rate constant of excited sensitizer quenching by BP were determined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1022 Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
C11H14O3