EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Co-sensitivity to disperse and reactive dyes.
PMID 19338592
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21620 Disperse Blue 124, for microscopy