EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Design and synthesis of N-2,6-difluorophenyl-5-methoxyl-1,2,4-triazolo[1,5-a]-pyrimidine-2-sulfonamide as acetohydroxyacid synthase inhibitor.
PMID 19342247
Abstract
Triazolopyrimidine-2-sulfonamide belongs to a herbicide group called acetohydroxyacid synthase inhibitors. With the aim to discover new triazolopyrimidine sulfonanilide compounds with high herbicidal activity and faster degradation rate in soil, the methyl group of Flumetsulam (FS) was modified into a methoxy group to produce a new herbicidal compound, N-2,6-difluorophenyl-5-methoxy-1,2,4-triazolo[1,5-a]pyrimidine-2-sulfonamide (experimental code: Y6610). The enzymatic kinetic results indicated that compound Y6610 and FS have k(i) values of 3.31x10(-6) M and 3.60x10(-7) M against Arabidopsis thaliana AHAS, respectively. The 10-fold lower enzyme-inhibiting activity of Y6610 was explained rationally by further computational simulations and binding free energy calculations. In addition, compound Y6610 was found to display the same level in vivo post-emergent herbicidal activity as FS against some broad-leaf weeds and good safety to rice, maize, and wheat at the dosages of 75-300 gai/ha. Further determination of the half-lives in soil revealed that the half-life in soil of Y6610 is 3.9 days shorter than that of FS. The experimental results herein showed that compound Y6610 could be regarded as a new potential acetohydroxyacid synthase-inhibiting herbicide candidate for further study.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196614
2,6-Difluoroaniline, ≥97%
C6H5F2N