EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Inositol 1,4,5-trisphosphate 3-kinase B is a negative regulator of BCR signaling that controls B cell selection and tolerance induction.
PMID 19342645
Abstract
Inositol 1,4,5-trisphosphate 3-kinase B (or Itpkb) converts inositol 1,4,5-trisphosphate to inositol 1,3,4,5-tetrakisphosphate upon Ag receptor activation and controls the fate and function of lymphocytes. To determine the role of Itpkb in B cell tolerance, Itpkb(-/-) mice were crossed to transgenic mice that express a BCR specific for hen egg lysozyme (IgHEL). B cells from Itpkb(-/-) IgHEL mice possess an anergic phenotype, hypoproliferate in response to cognate Ag, and yet they exhibit enhanced Ag-induced calcium signaling. In IgHEL transgenic mice that also express soluble HEL, lack of Itpkb converts anergy induction to deletion. These data establish Itpkb as a negative regulator of BCR signaling that controls the fate of developing B cells and tolerance induction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I8636
D-myo-Inositol 1,3,4,5-tetrakis(phosphate) ammonium salt, ≥95%
C6H16O18P4·8H3N