EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Metabolic engineering of Saccharomyces cerevisiae for production of novel cyanophycins with an extended range of constituent amino acids.
PMID 19346356