EMAIL THIS PAGE TO A FRIEND
Free radical research
Elevated neuronal nitric oxide synthase expression during ageing and mitochondrial energy production.
PMID 19347761
Abstract
This study evaluated the effect of ageing on brain mitochondrial function mediated through protein post-translational modifications. Neuronal nitric oxide synthase increased with age and this led to a discreet pattern of nitration of mitochondrial proteins. LC/MS/MS analyses identified the nitrated mitochondrial proteins as succinyl-CoA-transferase and F1-ATPase; the latter was nitrated at Tyr269, suggesting deficient ADP binding to the active site. Activities of succinyl-CoA-transferase, F1-ATPase and cytochrome oxidase decreased with age. The decreased activity of the latter cannot be ascribed to protein modifications and is most likely due to a decreased expression and assembly of complex IV. Mitochondrial protein post-translational modifications were associated with a moderately impaired mitochondrial function, as indicated by the decreased respiratory control ratios as a function of age and by the release of mitochondrial cytochrome c to the cytosol, thus supporting the amplification of apoptotic cascades.