EMAIL THIS PAGE TO A FRIEND
The Journal of Comparative Neurology
Variability in the occurrence of nitric oxide synthase immunoreactivity in different populations of rat sympathetic preganglionic neurons.
PMID 19350665
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AG220 Choline Acetyltransferase, for Blocking AB143, AB144P Antibodies