EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Activity profiles of cholinergic and intermingled GABAergic and putative glutamatergic neurons in the pontomesencephalic tegmentum of urethane-anesthetized rats.
PMID 19357291