EMAIL THIS PAGE TO A FRIEND
British dental journal
The role of phenoxymethylpenicillin, amoxicillin, metronidazole and clindamycin in the management of acute dentoalveolar abscesses--a review.
PMID 19357666
Abstract
Antibiotics are the most widely prescribed category of drugs issued on prescription by general dental practitioners. Despite this there remains little evidence-based literature on what should be prescribed for any given clinical situation, at what dosage and for how long. Given the current climate of evidence-based research, the need to keep antibiotic prescribing to an acceptable minimum, increasing levels of resistance of micro-organisms and widespread hospital infections with 'superbugs', there is a distinct need for appropriate prescribing guidelines. Considering best practice, an extensive review of the literature and a thorough understanding of current empirical treatment regimes, an attempt has been made to recommend suitable antibiotic prescribing for the adult patient suffering from acute dentoalveolar infections based on evidence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1000000
Phenoxymethylpenicillin, European Pharmacopoeia (EP) Reference Standard
C16H18N2O5S