EMAIL THIS PAGE TO A FRIEND
Talanta
Investigation of voltammetric enzyme-linked immunoassay system based on N-heterocyclic substrate of 2,3-diaminopyridine.
PMID 19362207
Abstract
A new voltammetric enzyme-linked immunoassay system using the electrochemical substrate 2,3-diaminopyridine (DAP) and horseradish peroxidase (HRP) system has been developed. DAP is oxidized with H2O2 catalyzed by HRP, and the resulting electroactive product produces a sensitive voltammetric peak at potential of -0.72V (vs. saturated calomel electrode (SCE)) in Britton-Robinson (BR) buffer solutions. The enzyme-catalyzed reaction conditions and voltammetric detection conditions have been investigated in detail. Under the selected optimum conditions, the linear range for detection of free HRP is from 6.0 x 10(-11) to 1.0 x 10(-8) g mL(-1) with a detection limit of 1.0 x 10(-12) g mL(-1). The new voltammetric detection system has been successfully applied for the assay of prostate specific antigen (PSA) in human serum ranging from 0.4 to 100 ng mL(-1) with a detection limit of 0.1 ng mL(-1), which is five times lower than that of traditional o-phenylenediamine (OPD) spectrophotometric enzyme-linked immunosorbent assay (ELISA) method. The proposed N-heterocyclic electrochemical detection system of DAP-H2O2-HRP has provided a new and much improved immunoassay method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125857
2,3-Diaminopyridine, 95%
C5H7N3