EMAIL THIS PAGE TO A FRIEND
Amino acids
(S)-alpha-methyl,alpha-amino acids: a new stereocontrolled synthesis.
PMID 19363669
Abstract
A new and convenient stereocontrolled synthesis of the optically pure (S)-alpha-methyl,alpha-amino acids 6(a-d) that exploits the chiral synthon 1,4-N,N-[(S)-1-phenylethyl]-piperazine-2,5-dione (1) is described. The (S)-1-phenylethyl group, bonded to each of the N-atoms of the 2,5-diketopiperazine, acts as a chiral inductor in the first alkylation, while the steric hindrance appears to be the determining factor of stereocontrol in third and forth alkylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
294357
DL-Homophenylalanine, 98%
C10H13NO2
544051
L-Homophenylalanine hydrochloride, 97%
C10H13NO2 · HCl