EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Identification and SAR of squarate inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).
PMID 19364658
Abstract
A novel series of inhibitors for mitogen activated protein kinase-activated protein kinase 2 (MK-2) are reported. These squarate based inhibitors were identified via a high-throughput screen. An MK2 co-structure with the starting ligand was obtained and a structure based approach was followed to optimize potency and selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123447
3,4-Dihydroxy-3-cyclobutene-1,2-dione, ≥99.0% (HPLC)
C4H2O4