EMAIL THIS PAGE TO A FRIEND
Nature
Biochemistry: Anchors away.
PMID 19370021
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3876
2′-Deoxyuridine 5′-monophosphate disodium salt, Sigma Grade
C9H11N2O8P · 2Na