EMAIL THIS PAGE TO A FRIEND
Amino acids
Synthesis of proline-derived dipeptides and their catalytic enantioselective direct aldol reactions: catalyst, solvent, additive and temperature effects.
PMID 19370392
Abstract
A series of dipeptides of L-proline-L-amino acid and L-proline-D-amino acid were synthesized to evaluate the catalytic effect for asymmetric direct aldol reactions. In the direct aldol reaction, a catalyst of L-proline-L-amino acid achieves better enantioselectivity than the corresponding L-proline-D-amino acid catalyst. Solubility of the dipeptide catalysts in the solvents is a key point for achieving a better yield of the direct aldol reaction, while hydrogen bonding of solvent does not play an important role in attaining better enantioselectivity and yield. Yield and enantioselectivity of the direct aldol reaction in water were improved by NMM and SDS additives, but the results that were done in plain DMSO were even better.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0880
Pro-Gly
C7H12N2O3