EMAIL THIS PAGE TO A FRIEND
Natural product communications
Antiangiogenic resveratrol analogues by mild m-CPBA aromatic hydroxylation of 3,5-dimethoxystilbenes.
PMID 19370931
Abstract
A mild treatment of the resveratrol analogue 3,5,4'-trimethoxystilbene 2 with m-CPBA afforded two hydroxylated methoxystilbenes 5 and 6 by direct aromatic hydroxylation. A similar protocol was applied to other stilbenes bearing a 3,5-dimethoxy moiety, namely tetramethoxystilbenes 7 and 10 to obtain respectively the hydroxylated analogues 8, 9 and 11, 12. The substrate 2 and the new compounds 5, 8 and 11 were evaluated as anti-angiogenic agents and proved significantly active in the range 1-100 microM.