EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antitubercular activity of novel 4-substituted imidazolyl-2,6-dimethyl-N3,N5-bisaryl-1,4-dihydropyridine-3,5-dicarboxamides.
PMID 19371979
Abstract
A series of 4-substituted imidazolyl-2,6-dimethyl-N(3),N(5)-bisaryl-1,4-dihydropyridine-3,5-dicarboxamides were prepared. They were screened as antitubercular agents against Mycobacterium tuberculosis H(37)Rv. Minimum inhibitory concentrations (MICs) were determined using agar proportion method. Compound 3i with 1-benzyl-2-methylthio-1H-imidazole-5-yl substituent at C-4 position and 4'-chloromophenyl group at C-3 and C-5 positions of the 1,4-dihydropyridine ring was the most potent one among the tested compounds. It was as potent as rifampicin against M. tuberculosis H(37)RV. Compound 3l also was an active antitubercular agent with the same substituent as compound 3i at the C-4 position and 3'-pyridyl group at C-3 and C-5 positions of the 1,4-dihydropyridine ring.