EMAIL THIS PAGE TO A FRIEND
Hepato-gastroenterology
Perhexiline maleate-induced hepatitis.
PMID 1937378
Abstract
We report on a case of perhexiline maleate-induced hepatitis secondary to a long-term administration of recommended daily dosages of 300 mg. The patient had a spectacular weight loss of 29 kg. He developed hepatitis, which subsided after drug withdrawal. Our electron-microscopic findings with the typical inclusion bodies and impaired hydroxylation capacity point to an underlying metabolic disorder as the pathogenetic mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0120
Perhexiline maleate salt, ≥98% (HPLC)
C19H35N · C4H4O4