EMAIL THIS PAGE TO A FRIEND
Molecular diversity
One hundred years of Meldrum's acid: advances in the synthesis of pyridine and pyrimidine derivatives.
PMID 19381852
Abstract
A general review (138 references) focused on the recent advances in the application of Meldrum's acid reactivity for synthesis of diverse pyridine and pyrimidine derivatives, mostly small and drug-like molecules is presented.