EMAIL THIS PAGE TO A FRIEND
Kinderkrankenschwester : Organ der Sektion Kinderkrankenpflege
[Study of skin care in premature and newborn infants].
PMID 19382435
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L7387 Lanolin
49909 Lanolin, tested according to Ph. Eur.