EMAIL THIS PAGE TO A FRIEND
Indian journal of pediatrics
'Glutaric aciduria type I--an easily diagnosable and treatable metabolic disorder'.
PMID 19390803
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89147
Glutaric acid, certified reference material, TraceCERT®
C5H8O4