EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Pancreatic beta cells express two autoantigenic forms of glutamic acid decarboxylase, a 65-kDa hydrophilic form and a 64-kDa amphiphilic form which can be both membrane-bound and soluble.
PMID 1939164