EMAIL THIS PAGE TO A FRIEND
The Journal of infectious diseases
Sialic acid: a preventable signal for pneumococcal biofilm formation, colonization, and invasion of the host.
PMID 19392624
Abstract
The correlation between carbohydrate availability, pneumococcal biofilm formation, nasopharyngeal colonization, and invasion of the host has been investigated. Of a series of sugars, only sialic acid (i.e., N-acetylneuraminic acid) enhanced pneumococcal biofilm formation in vitro, at concentrations similar to those of free sialic acid in human saliva. In a murine model of pneumococcal carriage, intranasal inoculation of sialic acid significantly increased pneumococcal counts in the nasopharynx and instigated translocation of pneumococci to the lungs. Competition of both sialic acid-dependent phenotypes was found to be successful when evaluated using the neuraminidase inhibitors DANA (i.e., 2,3-didehydro-2-deoxy-N-acetylneuraminic acid), zanamivir, and oseltamivir. The association between levels of free sialic acid on mucosae, pneumococcal colonization, and development of invasive disease shows how a host-derived molecule can influence a colonizing microbe and also highlights a molecular mechanism that explains the epidemiologic correlation between respiratory infections due to neuraminidase-bearing viruses and bacterial pneumonia. The data provide a new paradigm for the role of a host compound in infectious diseases and point to new treatment strategies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9050
N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid, ≥93%
C11H17NO8