EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A protease inhibitor discovery method using fluorescence correlation spectroscopy with position-specific labeled protein substrates.
PMID 19394304
Abstract
We developed novel substrates for protease activity evaluation by fluorescence correlation spectroscopy (FCS). Substrates were labeled in a position-specific manner with a fluorophore near the N terminus and included a C-terminal, 30 kDa, highly soluble protein (elongation factor Ts [EF-Ts]). The C-terminal protein enhanced the substrate peptide solubility and increased the molecular weight, enabling sensitive detection by FCS. Using the labeled substrates, caspase-3 and matrix metalloproteinase-9 (MMP-9) activities were confirmed by FCS. To demonstrate the suitability of this FCS-based assay for high-throughput screening, we screened various chemical compounds for MMP-9 inhibitors. The screening results confirmed the inhibitory activity of one compound and also revealed another potential MMP-9 inhibitor. Thus, this combination of position-specific labeled protein substrates and FCS may serve as a useful tool for evaluating activities of various proteases and for protease inhibitor screening.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6671
Actinonin
C19H35N3O5