EMAIL THIS PAGE TO A FRIEND
International journal of antimicrobial agents
Re: Serenoa repens associated with Urtica dioica (ProstaMEV) and curcumin and quercitin (FlogMEV) extracts are able to improve the efficacy of prulifloxacin in bacterial prostatitis patients: results from a prospective randomised study.
PMID 19394800