EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Palladium(0)-catalyzed cross-coupling of potassium (Z)-2-chloroalk-1-enyl trifluoroborates: a chemo- and stereoselective access to (Z)-chloroolefins and trisubstituted alkenes.
PMID 19396890
Abstract
We describe the preparation of a series of new potassium trifluoroborates 1 and the study of their behaviour in a Pd(0)-catalyzed cross-coupling reaction. We found that compounds 1 are endowed with original properties as they behave as nucleophilic cross-coupling partners chemoselectively but also as ambivalent synthons. The usefulness of this methodology has been successfully illustrated by the first total synthesis of an N-acyl spermidine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
768103
Dichlorobis(di-tert-butylphenylphosphine)palladium(II), 95%
C28H46Cl2P2Pd