EMAIL THIS PAGE TO A FRIEND
Journal of clinical pharmacology
Clopidogrel inhibits CYP2C19-dependent hydroxylation of omeprazole related to CYP2C19 genetic polymorphisms.
PMID 19398604
Abstract
This study explores the impact of clopidogrel on the pharmacokinetics of omeprazole related to CYP2C19 genetic polymorphisms. Twelve healthy volunteers (6 CYP2C19*1/*1, 5 CYP2C19*2/*2, and 1 CYP2C19*2/*3) are enrolled in a 2-phase randomized crossover trial. In each phase, the volunteers are administered a single oral dose of omeprazole 40 mg after pretreatment of either placebo or clopidogrel (300 mg on the first day and then 75 mg once daily for 3 consecutive days). Plasma concentrations of omeprazole and its metabolites are quantified by high-performance liquid chromatography with UV detection. After clopidogrel treatment, the AUC(0-infinity) of omeprazole increases by 30.02% +/- 18.03% (P = .004) and that of 5-hydroxyomeprazole decreases by 24.30% +/- 11.66% (P = .032) in CYP2C19*1/*1. The AUC(0-infinity) ratios of omeprazole to 5-hydroxyomeprazole increase by 74.98% +/- 35.48% (P = .001) and those of omeprazole to omeprazole sulfone do not change significantly (P = .832) in CYP2C19*1/*1. No significant alteration is observed in CYP2C19*2/*2 or *3. Clopidogrel inhibits CYP2C19-dependent hydroxylation of omeprazole in CYP2C19*1/*1 and has no impact on CYP3A4-catalyzed sulfoxidation of omeprazole.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
43999
5-Hydroxyomeprazole, analytical standard
C17H19N3O4S