EMAIL THIS PAGE TO A FRIEND
Journal of Korean medical science
Cessation of gonadotropin-releasing hormone antagonist on triggering day: an alternative method for flexible multiple-dose protocol.
PMID 19399268
Abstract
This study was performed to analyze retrospectively outcomes of stimulated in vitro fertilization (IVF) cycles where the gonadotropin-releasing hormone (GnRH) antagonist was omitted on ovulation triggering day. A total of 92 consecutive IVF cycles were included in 65 women who are undergoing ovarian stimulation with recombinant FSH. A GnRH antagonist, cetrorelix 0.25 mg/day, was started when leading follicle reached 14 mm in diameter until the day of hCG administration (Group A, 66 cycles) or until the day before hCG administration (Group B, 26 cycles). The duration of ovarian stimulation, total dose of gonadotropins, serum estradiol levels on hCG administration day, and the number of oocytes retrieved were not significantly different between the two groups. The total dose of GnRH antagonist was significantly lower in Group B compared to Group A (2.7+/-0.8 vs. 3.2+/-0.9 ampoules). There was no premature luteinization in the subjects. The proportion of mature oocytes (71.4% vs. 61.7%) and fertilization rate of mature (86.3+/-19.7% vs. 71.8+/-31.7%) was significantly higher in Group B. There were no significant differences in embryo quality and clinical pregnancy rates. Our results suggest that cessation of the GnRH antagonist on the day of hCG administration during a flexible multiple-dose protocol could reduce the total dose of GnRH antagonist without compromising IVF results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5249
Cetrorelix acetate, ≥98% (HPLC)
C70H92ClN17O14 · xC2H4O2