EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Nervous and vestibular toxicities of acrylonitrile and iminodipropionitrile.
PMID 19403855
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317306
3,3′-Iminodipropionitrile, technical grade, 90%
C6H9N3