EMAIL THIS PAGE TO A FRIEND
Gastroenterology
Treatment of hyperammonemia in liver failure: a tale of two enzymes.
PMID 19409290
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7125
L-Ornithine L-aspartate salt, powder
C9H19N3O6