EMAIL THIS PAGE TO A FRIEND
Molecular and Cellular Neurosciences
Brain-derived neurotrophic factor regulates early postnatal developmental cell death of dopamine neurons of the substantia nigra in vivo.
PMID 19409492