EMAIL THIS PAGE TO A FRIEND
Journal of endocrinological investigation
Clinical dilemmas arising from the increased intake of iodine in the Spanish population and the recommendation for systematic prescription of potassium iodide in pregnant and lactating women (Consensus of the TDY Working Group of SEEN).
PMID 19411820