EMAIL THIS PAGE TO A FRIEND
Anaesthesia
Myocardial infarction associated with the administration of intravenous ephedrine and metaraminol for spinal-induced hypotension.
PMID 19413828
Abstract
A 31-year-old female with no risk factors for cardiac disease suffered a peri-operative myocardial infarction during an elective gynaecological procedure under spinal anaesthesia. The timing and nature of cardiac symptoms suggest that the myocardial infarction was caused by coronary artery vasospasm secondary to ephedrine and/or metaraminol, which were administered to treat spinal-induced hypotension. We review the recent literature and case reports on myocardial infarction attributed to sympathomimetic drugs, and recommend the use of sublingual or intravenous nitrates when signs or symptoms of coronary arterial vasospasm become evident during their use.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4773
Metaraminol (+)-bitartrate salt
C9H13NO2 · C4H6O6