EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Spectrophotometric determination of hydralazine with 2-hydroxy-1-naphthaldehyde in pharmaceuticals.
PMID 1941570
Abstract
A new extraction-spectrophotometric method for the determination of hydralazine, based on its reaction with 2-hydroxy-1-naphthaldehyde at 25 degrees C, is described. The calibration curve was linear between 0.4 and 6 mg/mL of hydralazine. The molar absorbtivity of the product at 408 nm is 40,900 L.mol-1.cm-1. The method described was applied to the analysis of hydralazine in pharmaceutical preparations containing reserpine, hydrochlorothiazide, bendrofluorthiazine, propranolol, and other substances. The agreement with the U.S.P. XXI method was satisfactory for tablets and injections, but not for pellets.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H45353
2-Hydroxy-1-naphthaldehyde, technical grade
C11H8O2