EMAIL THIS PAGE TO A FRIEND
Toxicological sciences : an official journal of the Society of Toxicology
Neurotoxicogenomic investigations to assess mechanisms of action of the munitions constituents RDX and 2,6-DNT in Northern bobwhite (Colinus virginianus).
PMID 19417177
Abstract
Munitions constituents (MCs) including hexahydro-1,3,5-trinitro-1,3,5-triazine (RDX), 2,4,6-trinitrotoluene (TNT), and TNT derivatives are recognized to elicit aberrant neuromuscular responses in many species. The onset of seizures resulting in death was observed in the avian model Northern bobwhite after oral dosing with RDX beginning at 8 mg/kg/day in subacute (14 days) exposures, whereas affective doses of the TNT derivative, 2,6-dinitrotoluene (2,6-DNT), caused gastrointestinal impacts, lethargy, and emaciation in subacute and subchronic (60 days) exposures. To assess and contrast the potential neurotoxicogenomic effects of these MCs, a Northern bobwhite microarray was developed consisting of 4119 complementary DNA (cDNA) features enriched for differentially-expressed brain transcripts from exposures to RDX and 2,6-DNT. RDX affected hundreds of genes in brain tissue, whereas 2,6-DNT affected few (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D200603
2,6-Dinitrotoluene, 98%
C7H6N2O4