EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Antimicrobial activity of Artemisia douglasiana and dehydroleucodine against Helicobacter pylori.
PMID 19422904
Abstract
The increasing resistance of Helicobacter pylori to antibiotics demands the search for novel compounds from plant based sources. Artemisia douglasiana Besser is widely used in Cuyo region (Argentina) as folk medicine for the treatment of gastric ailments. Based on our previous studies that Artemisia douglasiana exert cytoprotective actions against ethanol-induced gastric mucosal injury we assayed the anti-Helicobacter pylori effect of the Artemisia douglasiana extract and its active compound, dehydroleucodine. The in vitro anti-bacterial activity of Artemisia douglasiana extract and its active compound, dehydroleucodine were determined against one standard strain and six clinical isolates of Helicobacter pylori by using the agar dilution methods. The results showed that both dehydroleucodine and Artemisia douglasiana extract had activity against the microorganism with MICs between 1-8 and 60-120 mg/L, respectively. Artemisia douglasiana may be a useful alternative treatment strategy principally in eradication of metronidazole and clarithromycin-resistant strain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4196
Dehydroleucodine, ≥98% (HPLC)
C15H16O3