EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Semisynthesis and pharmacological activities of Tetrac analogs: angiogenesis modulators.
PMID 19427201
Abstract
Novel Tetrac analogs were synthesized and then tested. Anti-angiogenesis efficacy was carried out using the Chick Chorioallantoic Membrane (CAM) model and the mouse matrigel model for angiogenesis. Pharmacological activities showed Tetrac can accommodate numerous modifications and maintain anti-angiogenesis activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3787
3,3′,5,5′-Tetraiodothyroacetic acid
C14H8I4O4