EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of penem inhibitors of bacterial signal peptidase.
PMID 19427205
Abstract
We report the first synthesis of a 5S penem, known to bind bacterial type I signal peptidase, from the commercially available and inexpensive 6-aminopenicillanic acid. We report the first in vivo activity of the compound and use structure-activity relationship studies to begin to define the determinants of signal peptidase binding and also to begin to optimize the penem as an antibiotic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A70909
(+)-6-Aminopenicillanic acid, 96%
C8H12N2O3S