EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Local examination of skin diffusion using FTIR spectroscopic imaging and multivariate target factor analysis.
PMID 19427483
Abstract
In the context of trans-dermal drug delivery it is very important to have mechanistic insight into the barrier function of the skin's stratum corneum and the diffusion mechanisms of topically applied drugs. Currently spectroscopic imaging techniques are evolving which enable a spatial examination of various types of samples in a dynamic way. ATR-FTIR imaging opens up the possibility to monitor spatial diffusion profiles across the stratum corneum of a skin sample. Multivariate data analyses methods based on factor analysis are able to provide insight into the large amount of spectroscopically complex and highly overlapping signals generated. Multivariate target factor analysis was used for spectral resolution and local diffusion profiles with time through stratum corneum. A model drug, 4-cyanophenol in polyethylene glycol 600 and water was studied. Results indicate that the average diffusion profiles between spatially different locations show similar profiles despite the heterogeneous nature of the biological sample and the challenging experimental set-up.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C94009
4-Cyanophenol, 95%
C7H5NO
45528
4-Hydroxybenzonitrile, PESTANAL®, analytical standard
C7H5NO