EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience methods
Intra-operatively obtained human tissue: protocols and techniques for the study of neural stem cells.
PMID 19427538
Abstract
The discoveries of neural (NSCs) and brain tumor stem cells (BTSCs) in the adult human brain and in brain tumors, respectively, have led to a new era in neuroscience research. These cells represent novel approaches to studying normal phenomena such as memory and learning, as well as pathological conditions such as Parkinson's disease, stroke, and brain tumors. This new paradigm stresses the importance of understanding how these cells behave in vitro and in vivo. It also stresses the need to use human-derived tissue to study human disease because animal models may not necessarily accurately replicate the processes that occur in humans. An important, but often underused, source of human tissue and, consequently, both NSCs and BTSCs, is the operating room. This study describes in detail both current and newly developed laboratory techniques, which in our experience are used to process and study human NSCs and BTSCs from tissue obtained directly from the operating room.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5726
Anti-Dlx2 Antibody, Anti-Dlx2 Antibody is an antibody against Dlx2 for use in WB, IH.
MAB3402
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 is an antibody against Glial Fibrillary Acidic Protein for use in IC, IH, IH(P) & WB with more than 55 product citations.
AB5622
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Detect Microtubule-Associated Protein 2 (MAP2) using this Anti-Microtubule-Associated Protein 2 (MAP2) Antibody validated for use in ELISA, IC, IH, IH(P) & WB with more than 55 product citations.
MAB5326
Anti-Nestin Antibody, clone 10C2, This Anti-Nestin Antibody, clone 10C2 is validated for use in IC, IH, IH(P), WB for the detection of Nestin.
MAB377
Anti-NeuN Antibody, clone A60, Anti-NeuN Antibody, clone A60 detects level of NeuN and has been published and validated for use in FC, IC, IF, IH, IH(P), IP and WB.
MAB345
Anti-O4 Antibody, clone 81, Anti-O4 Antibody, clone 81 is an antibody against O4 for use in IC, IH with more than 50 product citations.
AB9610
Anti-Olig-2 Antibody, Anti-Olig-2 Antibody is an antibody against Oligodendrocyte transcription factor 2 for use in IC, IH, IH(P), IP and WB.
MAB1637
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), Anti-Tubulin Antibody, beta III isoform, C-terminus, clone TU-20 (Similar to TUJ1) is an antibody against Tubulin for use in ELISA, IC, IH, IH(P), IP & WB with more than 55 product citations.
MAB3400
Anti-Vimentin Antibody, clone V9, Anti-Vimentin Antibody, clone V9 is an antibody against Vimentin for use in IC, IH, IH(P) & WB.
HPA015768
Anti-S100B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S9763
Sodium phosphate dibasic, ACS reagent, ≥99.0%
HNa2O4P
71640
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
HNa2O4P
04276
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
HNa2O4P